This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other POMT1 SIRNA Assays ›
Gene Symbol:
Pomt1
Gene Name:
protein-O-mannosyltransferase 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 11253424 - 11271873 on Build Rnor_6.0
UniGene ID:
Rn.162659
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136606

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pomt1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053406.2 NP_445858.1 8 844
XM_006234001.2 XP_006234063.1 8 1446
XM_008761776.1 XP_008759998.1 6 554
GenBank mRNA AF192388.1 - 8 817
BC070912.1 - 8 814 7104668

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145145, AY007238, AK153984, AK134770, BC027325, and others.

Gene Ontology Categories:

Function(s) Process(es)

extracellular matrix organization
protein O-linked mannosylation
dolichyl-phosphate-mannose-protein mannosyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0