This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SMPX SIRNA Assays ›
Gene Symbol:
Smpx
Gene Name:
small muscle protein, X-linked
Gene Aliases:
-
Chromosome Location:
Chr.X: 40030244 - 40086870 on Build Rnor_6.0
UniGene ID:
Rn.4123
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136583

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Smpx
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053395.1 NP_445847.1 5 758
XM_006256945.2 XP_006257007.1 5 727
GenBank mRNA AF364071.1 - 5 758

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header