This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD40LG SIRNA Assays ›
Gene Symbol:
Cd40lg
Gene Name:
CD40 ligand
Gene Aliases:
Cd40l, Tnfsf5
Chromosome Location:
Chr.X: 159703703 - 159714886 on Build Rnor_6.0
UniGene ID:
Rn.44218
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136446

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd40lg
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053353.1 NP_445805.1 2 188
GenBank mRNA AF013985.1 - 2 188
AF116582.1 - 2 188
EF066490.1 - 2 188

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of immune response to tumor cell
inflammatory response
immune response
platelet activation
B cell differentiation
positive regulation of B cell proliferation
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
B cell proliferation
negative regulation of apoptotic process
isotype switching
positive regulation of MHC class II biosynthetic process
immunoglobulin secretion
regulation of immunoglobulin secretion
dendritic cell differentiation
positive regulation of endothelial cell apoptotic process
molecular_function
cytokine activity
tumor necrosis factor receptor binding
CD40 receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0