This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SYNJ2 SIRNA Assays ›
Gene Symbol:
Synj2
Gene Name:
synaptojanin 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 46835922 - 46932544 on Build Rnor_6.0
UniGene ID:
Rn.10868
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136392

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Synj2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113371.1 NP_001106842.1 16 2249
NM_001113372.1 NP_001106843.1 16 2249
NM_032071.2 NP_114460.1 16 2304
GenBank mRNA AY034050.1 - 16 2249
AY034051.1 - 16 2249
U90312.1 - 16 2304

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

brain development
dephosphorylation
inositol phosphate dephosphorylation
phosphatidylinositol dephosphorylation
intracellular distribution of mitochondria
nucleotide binding
nucleic acid binding
RNA binding
phosphatidylinositol-4,5-bisphosphate 5-phosphatase activity
inositol-polyphosphate 5-phosphatase activity
protein binding
SH3 domain binding
PDZ domain binding
phosphoric ester hydrolase activity
phosphatidylinositol monophosphate phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm