This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC12A2 SIRNA Assays ›
Gene Symbol:
Slc12a2
Gene Name:
solute carrier family 12 (sodium/potassium/chloride transporter), member 2
Gene Aliases:
Bsc2, Nkcc1
Chromosome Location:
Chr.18: 52917124 - 52985281 on Build Rnor_6.0
UniGene ID:
Rn.11523
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136150

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc12a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031798.1 NP_113986.1 23 3227
XM_006254752.2 XP_006254814.1 22 3110
GenBank mRNA AF051561.1 - 23 3227
AF071863.1 - 23 2510
FQ211948.1 - 7 591

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
sodium ion transport
chloride transport
hyperosmotic response
gamma-aminobutyric acid signaling pathway
aging
ammonium transport
transepithelial chloride transport
multicellular organism growth
positive regulation of cell volume
detection of mechanical stimulus involved in sensory perception of sound
branching involved in mammary gland duct morphogenesis
mammary duct terminal end bud growth
transepithelial ammonium transport
ammonium transmembrane transport
chloride transmembrane transport
sodium:potassium:chloride symporter activity
ammonium transmembrane transporter activity
cation:chloride symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0