This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other POLR1A SIRNA Assays ›
Gene Symbol:
Polr1a
Gene Name:
polymerase (RNA) I polypeptide A
Gene Aliases:
Rpa1, Rpo1-4
Chromosome Location:
Chr.4: 99822968 - 99896052 on Build Rnor_6.0
UniGene ID:
Rn.24877
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136077

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Polr1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031772.1 NP_113960.1 6 696
XM_008762998.1 XP_008761220.1 6 973
GenBank mRNA AF025425.1 - 6 696

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase I promoter
rRNA transcription
RNA polymerase I activity
DNA binding
DNA-directed RNA polymerase activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0