This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDXK SIRNA Assays ›
Gene Symbol:
Pdxk
Gene Name:
pyridoxal (pyridoxine, vitamin B6) kinase
Gene Aliases:
-
Chromosome Location:
Chr.20: 10930651 - 10952194 on Build Rnor_6.0
UniGene ID:
Rn.18815
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136068

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pdxk
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031769.1 NP_113957.1 3, 4 248
GenBank mRNA AF020346.1 - 3, 4 248
FQ213832.1 - 3, 4 352

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172134, AK138248, AK080846, AK166464, AK145470, and others.

Gene Ontology Categories:

Function(s) Process(es)

cell proliferation
pyridoxal 5'-phosphate salvage
response to toxic substance
response to X-ray
response to amine
phosphorylation
response to insecticide
response to food
response to progesterone
response to drug
pyridoxal phosphate biosynthetic process
negative regulation of apoptotic process
magnesium ion binding
ATP binding
drug binding
zinc ion binding
pyridoxal kinase activity
pyridoxal phosphate binding
potassium ion binding
sodium ion binding
lithium ion binding
protein homodimerization activity
metal ion binding
pyridoxal binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0