This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SHARPIN SIRNA Assays ›
Gene Symbol:
Sharpin
Gene Name:
SHANK-associated RH domain interactor
Gene Aliases:
Conneck1
Chromosome Location:
Chr.7: 117413151 - 117417363 on Build Rnor_6.0
UniGene ID:
Rn.38393
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135854

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sharpin
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031153.2 NP_112415.1 7 1286
GenBank mRNA AF203906.1 - 7 960
BC083553.1 - 7 1286 7189111
FQ213170.1 - 7 904

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitochondrion organization
brain development
epidermis development
regulation of tumor necrosis factor-mediated signaling pathway
apoptotic nuclear changes
keratinization
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of inflammatory response
protein homooligomerization
protein linear polyubiquitination
regulation of CD40 signaling pathway
protein binding
zinc ion binding
polyubiquitin binding
protein complex binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0