This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TPP2 SIRNA Assays ›
Gene Symbol:
Tpp2
Gene Name:
tripeptidyl peptidase II
Gene Aliases:
TPP-2
Chromosome Location:
Chr.9: 50580779 - 50628942 on Build Rnor_6.0
UniGene ID:
Rn.28991
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135806

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tpp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031137.1 NP_112399.1 404
XM_006244852.2 XP_006244914.1 859
XM_006244853.2 XP_006244915.1 859
XM_006244870.2 XP_006244932.1 3, 4 859
GenBank mRNA U50194.1 - 404

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_003291, M73047, AK304313, BC039905, M55169, and others.
This siRNA also targets the following transcripts in Mouse: NM_009418, AK171519, AK154144, BC011275, AK038041, and others.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
aminopeptidase activity
serine-type endopeptidase activity
tripeptidyl-peptidase activity
peptide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0