This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other S100A10 SIRNA Assays ›
Gene Symbol:
S100a10
Gene Name:
S100 calcium binding protein A10
Gene Aliases:
p11
Chromosome Location:
Chr.2: 193892589 - 193901236 on Build Rnor_6.0
UniGene ID:
Rn.4083
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135737

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: S100a10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031114.1 NP_112376.1 2 148
GenBank mRNA AB453238.1 - 1 103
BC091565.1 - 2 174 7441806
FQ222734.1 - 2 203
FQ222778.1 - 2 171
FQ223502.1 - 2 168
FQ228469.1 - 2 171
FQ228745.1 - 2 171
FQ229216.1 - 2 171
J03627.1 - 2 148

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
membrane raft assembly
membrane budding
response to antidepressant
regulation of cell differentiation
positive regulation of binding
protein heterotetramerization
positive regulation of stress fiber assembly
positive regulation of focal adhesion assembly
establishment of protein localization to plasma membrane
positive regulation of substrate adhesion-dependent cell spreading
calcium ion binding
protein binding
lipid binding
protein homodimerization activity
ion channel binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0