This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MMP2 SIRNA Assays ›
Gene Symbol:
Mmp2
Gene Name:
matrix metallopeptidase 2
Gene Aliases:
-
Chromosome Location:
Chr.19: 15542771 - 15570589 on Build Rnor_6.0
UniGene ID:
Rn.6422
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135546

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mmp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031054.2 NP_112316.2 2, 3 673
GenBank mRNA BC074013.1 - 2, 3 603 7100171
U65656.1 - 2, 3 673

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008610, BC070430, M84324, AK148184.

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
ovarian follicle development
ovulation from ovarian follicle
luteinization
response to hypoxia
blood vessel maturation
intramembranous ossification
proteolysis
response to oxidative stress
negative regulation of cell adhesion
female pregnancy
embryo implantation
parturition
aging
response to radiation
response to mechanical stimulus
peripheral nervous system axon regeneration
response to activity
cell migration
positive regulation of cell migration
collagen catabolic process
response to retinoic acid
response to cytokine
response to nicotine
endodermal cell differentiation
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
response to estrogen
positive regulation of innate immune response
negative regulation of vasoconstriction
skeletal system morphogenesis
tissue remodeling
response to electrical stimulus
response to hyperoxia
face morphogenesis
bone trabecula formation
prostate gland epithelium morphogenesis
cellular response to amino acid stimulus
cellular response to cytokine stimulus
cellular response to interleukin-1
cellular response to estradiol stimulus
cellular response to fluid shear stress
fibronectin binding
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0