siRNA Details
Gene Symbol: Itpka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031045.2 | NP_112307.2 | 3, 4 | 850 | ||
GenBank mRNA | M29787.1 | - | 3, 4 | 850 | ||
X56917.1 | - | 3, 4 | 841 |

Gene Symbol: Itpka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031045.2 | NP_112307.2 | 3, 4 | 850 | ||
GenBank mRNA | M29787.1 | - | 3, 4 | 850 | ||
X56917.1 | - | 3, 4 | 841 |