This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPT SIRNA Assays ›
Gene Symbol:
Gpt
Gene Name:
glutamic-pyruvate transaminase (alanine aminotransferase)
Gene Aliases:
Gpt1
Chromosome Location:
Chr.7: 117757345 - 117761954 on Build Rnor_6.0
UniGene ID:
Rn.6318
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135504

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gpt
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031039.1 NP_112301.1 4 565
XM_008765599.1 XP_008763821.1 5 1285
GenBank mRNA BC085728.1 - 3 620
BC097937.1 - 4 569 7324977
D10354.1 - 4 565

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
biosynthetic process
L-alanine catabolic process
L-alanine:2-oxoglutarate aminotransferase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0