This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GNAI2 SIRNA Assays ›
Gene Symbol:
Gnai2
Gene Name:
guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2
Gene Aliases:
Galphai2
Chromosome Location:
Chr.8: 116370730 - 116391337 on Build Rnor_6.0
UniGene ID:
Rn.3036
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135493

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gnai2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031035.3 NP_112297.1 5 653
XM_006243858.1 XP_006243920.1 5 683
GenBank mRNA AY899210.1 - 5 501
BC078806.1 - 5 605 7121183
CO394955.1 - 5 653
DQ120471.1 - 5 562
DQ120472.1 - 5 562
M12672.1 - 5 549
M17528.1 - 5 651

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKK activity
adenosine receptor signaling pathway
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
G-protein coupled acetylcholine receptor signaling pathway
gamma-aminobutyric acid signaling pathway
G-protein coupled glutamate receptor signaling pathway
regulation of heart contraction
metabolic process
cell proliferation
positive regulation of cell proliferation
intracellular signal transduction
negative regulation of synaptic transmission
cell division
regulation of calcium ion transport
G-protein coupled receptor binding
GTPase activity
signal transducer activity
protein binding
GTP binding
G-protein beta/gamma-subunit complex binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0