This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANPEP SIRNA Assays ›
Gene Symbol:
Anpep
Gene Name:
alanyl (membrane) aminopeptidase
Gene Aliases:
AP-M, AP-N, Apm, Apn, KZP, Lap1, rAPN
Chromosome Location:
Chr.1: 141561364 - 141604249 on Build Rnor_6.0
UniGene ID:
Rn.11132
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135423

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Anpep
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031012.1 NP_112274.1 1 244
XM_006229407.2 XP_006229469.1 2 664
XM_006229408.2 XP_006229470.1 2 685
XM_006229409.1 XP_006229471.1 2 721
GenBank mRNA M25073.1 - 1 244
M26710.1 - 1 291

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008486, AK171427, BC005431, AK143746, U77083.

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
proteolysis
cellular aromatic compound metabolic process
cell differentiation
negative regulation of renal sodium excretion
aminopeptidase activity
metallopeptidase activity
zinc ion binding
peptide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0