This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRIM23 SIRNA Assays ›
Gene Symbol:
Trim23
Gene Name:
tripartite motif-containing 23
Gene Aliases:
Ard1, Arfd1
Chromosome Location:
Chr.2: 34255305 - 34288140 on Build Rnor_6.0
UniGene ID:
Rn.6763
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135288

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trim23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100637.1 NP_001094107.1 8 1350
XM_008760683.1 XP_008758905.1 9 1431
XM_008760684.1 XP_008758906.1 9 1332
GenBank mRNA L04760.1 - 8 1201

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ADP-ribosylation
small GTPase mediated signal transduction
protein ubiquitination
GTPase activity
ubiquitin-protein transferase activity
GTP binding
zinc ion binding
ligase activity
GDP binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0