This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC16A3 SIRNA Assays ›
Gene Symbol:
Slc16a3
Gene Name:
solute carrier family 16 (monocarboxylate transporter), member 3
Gene Aliases:
MCT4, Mct3
Chromosome Location:
Chr.10: 110138523 - 110148360 on Build Rnor_6.0
UniGene ID:
Rn.10826
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135270

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc16a3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030834.1 NP_110461.1 3 426
XM_006247931.2 XP_006247993.1 4 541
XM_006247932.2 XP_006247994.1 4 833
XM_006247933.2 XP_006247995.1 3 448
XM_006247934.2 XP_006247996.1 4 507
XM_008768513.1 XP_008766735.1 4 3265
XM_008768514.1 XP_008766736.1 3 3172
GenBank mRNA BC168146.1 - 3 385 9089640
U87627.1 - 3 426

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001038653, NM_030696, NM_001038654, AK165596, AK141381, and others.

Gene Ontology Categories:

Function(s) Process(es)

plasma membrane lactate transport
lactate transmembrane transporter activity
symporter activity
secondary active monocarboxylate transmembrane transporter activity
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0