This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHRNA1 SIRNA Assays ›
Gene Symbol:
Chrna1
Gene Name:
cholinergic receptor, nicotinic, alpha 1 (muscle)
Gene Aliases:
-
Chromosome Location:
Chr.3: 60445657 - 60460724 on Build Rnor_6.0
UniGene ID:
Rn.44633
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135223

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chrna1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024485.1 NP_077811.1 8 1148
GenBank mRNA X74832.1 - 8 1148

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

skeletal muscle contraction
cation transport
neuromuscular synaptic transmission
neuromuscular junction development
neuronal action potential
regulation of membrane potential
muscle cell cellular homeostasis
skeletal muscle tissue growth
musculoskeletal movement
neuromuscular process
neuron cellular homeostasis
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
acetylcholine receptor activity
acetylcholine binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0