This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other THOC6 SIRNA Assays ›
Gene Symbol:
Thoc6
Gene Name:
THO complex 6 homolog (Drosophila)
Gene Aliases:
Pdrp
Chromosome Location:
Chr.10: 12989135 - 12994495 on Build Rnor_6.0
UniGene ID:
Rn.42184
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135117

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Thoc6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024384.2 NP_077360.2 6, 7 658
GenBank mRNA BC079149.1 - 6, 7 658 7128541

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
mRNA export from nucleus
apoptotic process
central nervous system development
RNA splicing
negative regulation of apoptotic process
viral mRNA export from host cell nucleus
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0