This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DAB2 SIRNA Assays ›
Gene Symbol:
Dab2
Gene Name:
disabled 2, mitogen-responsive phosphoprotein
Gene Aliases:
Doc-2, Doc2
Chromosome Location:
Chr.2: 55747353 - 55768848 on Build Rnor_6.0
UniGene ID:
Rn.161784
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135053

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dab2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024159.1 NP_077073.1 9 1172
GenBank mRNA BC097314.1 - 9 1207 7444786
U95177.1 - 9 1172

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023118, AK154716, CB598046, U18869.

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis involved in differentiation
in utero embryonic development
positive regulation of receptor recycling
positive regulation of protein phosphorylation
endocytosis
receptor-mediated endocytosis
pinocytosis
apoptotic process
integrin-mediated signaling pathway
activation of JUN kinase activity
endoderm development
excretion
positive regulation of epithelial to mesenchymal transition
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of neuron projection development
protein transport
Wnt signaling pathway
myeloid cell differentiation
negative regulation of cell growth
positive regulation of cell migration
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of protein binding
positive regulation of aldosterone biosynthetic process
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
positive regulation of transcription elongation from RNA polymerase II promoter
leading edge cell differentiation
negative regulation of apoptotic process
positive regulation of cell adhesion
positive regulation of endocytosis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
clathrin coat assembly
response to steroid hormone
negative regulation of epithelial cell proliferation
positive regulation of SMAD protein import into nucleus
negative regulation of androgen receptor signaling pathway
negative regulation of ERK1 and ERK2 cascade
cellular response to epidermal growth factor stimulus
cellular response to transforming growth factor beta stimulus
negative regulation of canonical Wnt signaling pathway
positive regulation of substrate adhesion-dependent cell spreading
response to salt
negative regulation of protein localization to plasma membrane
positive regulation of Wnt signaling pathway, planar cell polarity pathway
regulation of Rho-dependent protein serine/threonine kinase activity
positive regulation of clathrin-mediated endocytosis
positive regulation of early endosome to late endosome transport
positive regulation of aldosterone secretion
positive regulation of integrin-mediated signaling pathway
negative regulation of extrinsic apoptotic signaling pathway
integrin binding
protein binding
phosphatidylinositol-4,5-bisphosphate binding
protein C-terminus binding
clathrin binding
phosphatidylinositol binding
AP-2 adaptor complex binding
clathrin adaptor activity
cargo receptor activity
SMAD binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0