This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANXA6 SIRNA Assays ›
Gene Symbol:
Anxa6
Gene Name:
annexin A6
Gene Aliases:
Anx6, CPB-II
Chromosome Location:
Chr.10: 40320581 - 40375605 on Build Rnor_6.0
UniGene ID:
Rn.204053
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135042

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Anxa6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024156.2 NP_077070.2 5 331
XM_006246340.2 XP_006246402.1 5 1069
XM_006246341.2 XP_006246403.1 5 1070
GenBank mRNA BC072523.1 - 5 280 7105847
CB585592.1 - 5 349

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

calcium ion transport
regulation of muscle contraction
ion transmembrane transport
secretion
protein homooligomerization
calcium ion binding
GTP binding
calcium-dependent phospholipid binding
lipid binding
ligand-gated ion channel activity
cholesterol binding
protein homodimerization activity
calcium-dependent protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0