This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARF4 SIRNA Assays ›
Gene Symbol:
Arf4
Gene Name:
ADP-ribosylation factor 4
Gene Aliases:
-
Chromosome Location:
Chr.16: 2338333 - 2354909 on Build Rnor_6.0
UniGene ID:
Rn.35935
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s135027

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arf4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024151.1 NP_077065.1 4 509
GenBank mRNA BC063167.1 - 4 467 6921397
FQ213125.1 - 4 492
FQ213804.1 - 4 506
FQ234083.1 - 4 467
FQ234248.1 - 4 579
L12383.1 - 4 509

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ADP-ribosylation
epidermal growth factor receptor signaling pathway
small GTPase mediated signal transduction
brain development
protein transport
vesicle-mediated transport
cell migration
activation of phospholipase D activity
negative regulation of apoptotic process
apical protein localization
establishment or maintenance of epithelial cell apical/basal polarity
positive regulation of transcription from RNA polymerase II promoter
response to axon injury
protein localization to cilium
regulation of reactive oxygen species metabolic process
epidermal growth factor receptor binding
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0