This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MRS2 SIRNA Assays ›
Gene Symbol:
Mrs2
Gene Name:
MRS2 magnesium transporter
Gene Aliases:
Mrs2l, Rpt
Chromosome Location:
Chr.17: 42064258 - 42083602 on Build Rnor_6.0
UniGene ID:
Rn.33647
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s134991

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mrs2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024001.1 NP_076491.1 6 721
XM_006253937.2 XP_006253999.1 6 839
GenBank mRNA AF288289.1 - 6 721
AF288290.1 - 2 172
AF288291.1 - 2 174
BC083554.1 - 6 714
BC098916.1 - 6 696

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

magnesium ion transport
magnesium ion transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0