This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNIP3 SIRNA Assays ›
Gene Symbol:
Kcnip3
Gene Name:
Kv channel interacting protein 3, calsenilin
Gene Aliases:
Csen, Dream, rKChIP3
Chromosome Location:
Chr.3: 120023062 - 120087157 on Build Rnor_6.0
UniGene ID:
Rn.74239
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s134769

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnip3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032462.2 NP_115851.1 6 563
XM_006234989.2 XP_006235051.1 6 692
XM_006234990.2 XP_006235052.1 5 4008
GenBank mRNA AB043892.1 - 5 542
AF297118.1 - 6 565

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
transcription from RNA polymerase II promoter
intracellular protein transport
apoptotic process
sensory perception of pain
regulation of neuron apoptotic process
response to pain
behavioral response to pain
potassium ion transmembrane transport
protein localization to plasma membrane
regulation of potassium ion transmembrane transport
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
voltage-gated ion channel activity
potassium channel activity
calcium ion binding
protein binding
protein C-terminus binding
potassium channel regulator activity
sequence-specific DNA binding
ion channel binding
calcium-dependent protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0