This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GABRQ SIRNA Assays ›
Gene Symbol:
Gabrq
Gene Name:
gamma-aminobutyric acid (GABA) A receptor, theta
Gene Aliases:
-
Chromosome Location:
Chr.X: 152852243 - 152865049 on Build Rnor_6.0
UniGene ID:
Rn.81067
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s134738

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gabrq
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031733.1 NP_113921.1 4 467
XM_006229516.2 XP_006229578.1 5 631
GenBank mRNA AF419333.1 - 4 467

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
ion transmembrane transport
GABA-A receptor activity
extracellular ligand-gated ion channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header