This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RUVBL1 SIRNA Assays ›
Gene Symbol:
Ruvbl1 RGD1564855
Gene Name:
RuvB-like AAA ATPase 1
similar to RuvB-like protein 1
Gene Aliases:
NMP238, Pontin52, Tip49a
-
Chromosome Location:
Chr.4: 120411917 - 120447458 on Build Rnor_6.0
Chromosome Location:
Chr.X: 1854580 - 1856333 on Build Rnor_6.0
UniGene ID:
Rn.86410
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s134640

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ruvbl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147177.1 NP_671706.1 1146
XM_006236865.2 XP_006236927.1 1220
GenBank mRNA AB001581.1 - 1133
AB002406.1 - 1146
BC072511.1 - 1131 7104777
BC086531.1 - 1164 7304521
Gene Symbol: RGD1564855
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773046.1 XP_008771268.1 1 1191

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019685, AK145062, AK012170, AF100694, AK132783, and others.

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA recombination
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
positive regulation of plasminogen activation
DNA duplex unwinding
regulation of growth
histone H4 acetylation
histone H2A acetylation
cell division
regulation of fibroblast apoptotic process
DNA helicase activity
ATP binding
ATPase activity
ATP-dependent 3'-5' DNA helicase activity
ATP-dependent 5'-3' DNA helicase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0