This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PIK3C3 SIRNA Assays ›
Gene Symbol:
Pik3c3
Gene Name:
phosphatidylinositol 3-kinase, catalytic subunit type 3
Gene Aliases:
-
Chromosome Location:
Chr.18: 22964129 - 23047896 on Build Rnor_6.0
UniGene ID:
Rn.30010
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s134601

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pik3c3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022958.2 NP_075247.1 6 726
XM_006254488.2 XP_006254550.1 6 807
GenBank mRNA AJ006710.1 - 6 708
BC061981.1 - 6 726 5597998

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

autophagosome assembly
protein phosphorylation
endosome organization
protein processing
phosphatidylinositol-3-phosphate biosynthetic process
response to leucine
early endosome to late endosome transport
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
regulation of protein secretion
protein kinase activity
protein binding
ATP binding
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol 3-kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0