This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLIT1 SIRNA Assays ›
Gene Symbol:
Slit1
Gene Name:
slit homolog 1 (Drosophila)
Gene Aliases:
MEGF4
Chromosome Location:
Chr.1: 260843481 - 260992366 on Build Rnor_6.0
UniGene ID:
Rn.30002
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s134587

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slit1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022953.2 NP_075242.1 17 1814
GenBank mRNA AB011530.1 - 17 1814
AB073214.1 - 17 1739
AB073215.1 - 17 1739
AF133730.1 - 17 1655

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

axonogenesis
axon guidance
brain development
motor neuron axon guidance
spinal cord development
olfactory bulb development
chemorepulsion involved in embryonic olfactory bulb interneuron precursor migration
tangential migration from the subventricular zone to the olfactory bulb
telencephalon cell migration
retinal ganglion cell axon guidance
dorsal/ventral axon guidance
establishment of nucleus localization
neuron projection morphogenesis
negative regulation of axon extension involved in axon guidance
axon extension involved in axon guidance
negative chemotaxis
negative regulation of synapse assembly
calcium ion binding
protein binding
heparan sulfate proteoglycan binding
Roundabout binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0