This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BCHE SIRNA Assays ›
Gene Symbol:
Bche
Gene Name:
butyrylcholinesterase
Gene Aliases:
-
Chromosome Location:
Chr.2: 171100137 - 171196186 on Build Rnor_6.0
UniGene ID:
Rn.48791
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s134555

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bche
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022942.1 NP_075231.1 2 539
XM_008761112.1 XP_008759334.1 1 616
GenBank mRNA AF244349.1 - 2 539

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to nutrient
learning
negative regulation of cell proliferation
neuroblast differentiation
choline metabolic process
response to drug
response to alkaloid
negative regulation of synaptic transmission
response to glucocorticoid
response to folic acid
acetylcholinesterase activity
cholinesterase activity
choline binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0