This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GOLGA2 SIRNA Assays ›
Gene Symbol:
Golga2
Gene Name:
golgin A2
Gene Aliases:
Gm130
Chromosome Location:
Chr.3: 11317328 - 11337569 on Build Rnor_6.0
UniGene ID:
Rn.6165
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s134206

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Golga2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022596.2 NP_072118.2 12 917
XM_006233992.2 XP_006234054.1 13 967
XM_006233993.2 XP_006234055.1 13 967
XM_006233994.2 XP_006234056.1 13 967
XM_006233995.2 XP_006234057.1 12 886
XM_006233996.2 XP_006234058.1 12 887
XM_006233997.2 XP_006234059.1 12 886
XM_006233998.2 XP_006234060.1 12 886
XM_008761774.1 XP_008759996.1 11 791
GenBank mRNA FQ147221.1 - 6 351
U35022.2 - 12 917

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
negative regulation of protein binding
positive regulation of axonogenesis
Golgi localization
protein binding
protein kinase binding
syntaxin binding
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0