This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ST3GAL3 SIRNA Assays ›
Gene Symbol:
St3gal3
Gene Name:
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Gene Aliases:
Siat6
Chromosome Location:
Chr.5: 136765309 - 136965642 on Build Rnor_6.0
UniGene ID:
Rn.44390
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s134082

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: St3gal3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031697.1 NP_113885.1 6 417
XM_006238715.2 XP_006238777.1 6 854
XM_006238716.2 XP_006238778.1 5 820
XM_008764066.1 XP_008762288.1 6 3344
XM_008764067.1 XP_008762289.1 6 437
GenBank mRNA M97754.1 - 6 417

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
sialylation
beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
N-acetyllactosaminide alpha-2,3-sialyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0