This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RPL24 SIRNA Assays ›
Gene Symbol:
Rpl24 RGD1560821
Gene Name:
ribosomal protein L24
similar to small nuclear ribonucleoparticle-associated protein
Gene Aliases:
-
-
Chromosome Location:
Chr.11: 47116885 - 47122093 on Build Rnor_6.0
Chromosome Location:
Chr.X: 77885562 - 77886179 on Build Rnor_6.0
UniGene ID:
Rn.118133
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s133943

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rpl24
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022515.1 NP_071960.1 3 184
GenBank mRNA BC058473.1 - 3 209 6918141
FQ209984.1 - 3 203
FQ220432.1 - 3 213
FQ221142.1 - 3 202
FQ221221.1 - 3 209
FQ221486.1 - 3 205
FQ221683.1 - 3 196
FQ221688.1 - 3 205
FQ221810.1 - 3 205
FQ221946.1 - 3 205
FQ221988.1 - 3 209
FQ222823.1 - 3 205
FQ223282.1 - 3 205
FQ224199.1 - 3 205
FQ224575.1 - 3 209
FQ224676.1 - 3 205
X78443.1 - 3 184
Gene Symbol: RGD1560821
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257178.2 XP_006257240.1 2 197

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ribosomal large subunit assembly
translation
mitotic cell cycle checkpoint
exit from mitosis
optic nerve development
retinal ganglion cell axon guidance
retina development in camera-type eye
structural constituent of ribosome
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0