This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CAPN10 SIRNA Assays ›
Gene Symbol:
Capn10
Gene Name:
calpain 10
Gene Aliases:
-
Chromosome Location:
Chr.9: 100104000 - 100112833 on Build Rnor_6.0
UniGene ID:
Rn.176334
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s133571

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Capn10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031673.2 NP_113861.1 1 288
GenBank mRNA AB113362.1 - 1 195
AB113363.1 - 1 195
AF227909.3 - 1 188
BC085725.1 - 1 288 7123652

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitophagy
proteolysis
mitochondrion organization
aging
response to glucose
actin cytoskeleton reorganization
response to nutrient levels
positive regulation of insulin secretion
positive regulation of intracellular transport
cellular response to insulin stimulus
positive regulation of apoptotic process
positive regulation of glucose import
type B pancreatic cell apoptotic process
positive regulation of type B pancreatic cell apoptotic process
SNARE binding
calcium-dependent cysteine-type endopeptidase activity
protein binding
cytoskeletal protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0