This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LIN7C SIRNA Assays ›
Gene Symbol:
Lin7c
Gene Name:
lin-7 homolog C (C. elegans)
Gene Aliases:
Veli3
Chromosome Location:
Chr.3: 101010923 - 101018235 on Build Rnor_6.0
UniGene ID:
Rn.9461
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s133406

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lin7c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021851.1 NP_068623.1 2 57
GenBank mRNA AF090136.1 - 2 57
FQ212254.1 - 89

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011699, BC052471, AF087695, AK128989, AF173083, and others.

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelial sheet
exocytosis
neurotransmitter secretion
asymmetric protein localization
protein transport
maintenance of epithelial cell apical/basal polarity
protein localization to basolateral plasma membrane
cytoskeletal protein binding
protein domain specific binding
PDZ domain binding
L27 domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm