This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SYNPO SIRNA Assays ›
Gene Symbol:
Synpo
Gene Name:
synaptopodin
Gene Aliases:
-
Chromosome Location:
Chr.18: 55870720 - 55891710 on Build Rnor_6.0
UniGene ID:
Rn.42910
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s133244

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Synpo
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021695.2 NP_067727.2 2 1650
GenBank mRNA AB013130.1 - 1 1176
AM980944.1 - 1120
DY313339.1 - 1 268

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of actin filament bundle assembly
regulation of stress fiber assembly
actin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0