This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MB SIRNA Assays ›
Gene Symbol:
Mb
Gene Name:
myoglobin
Gene Aliases:
-
Chromosome Location:
Chr.7: 118101633 - 118108864 on Build Rnor_6.0
UniGene ID:
Rn.40511
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s133117

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021588.2 NP_067599.1 2, 3 351
GenBank mRNA AF197916.1 - 2, 3 324
BC070511.1 - 2, 3 351 7103504
FQ214690.1 - 2, 3 383
FQ214898.1 - 2, 3 382
FQ214986.1 - 2, 3 383
FQ215026.1 - 2, 3 383
FQ215032.1 - 2, 3 382
FQ215043.1 - 2, 3 382
FQ215217.1 - 2, 3 381
FQ215441.1 - 2, 3 382
FQ215511.1 - 2, 3 381
FQ215553.1 - 2, 3 382
FQ215564.1 - 2, 3 381
FQ215642.1 - 2, 3 392
FQ215802.1 - 2, 3 383
FQ215880.1 - 2, 3 383
FQ215982.1 - 2, 3 382
FQ216001.1 - 2, 3 381
FQ216029.1 - 2, 3 380
FQ216127.1 - 2, 3 380
FQ216280.1 - 2, 3 389
FQ216299.1 - 2, 3 383
FQ216306.1 - 2, 3 384
FQ216335.1 - 2, 3 383
FQ216380.1 - 2, 3 382
FQ216443.1 - 2, 3 381
FQ216504.1 - 2, 3 382
FQ216709.1 - 2, 3 382
FQ216731.1 - 2, 3 381
FQ217450.1 - 2, 3 382
FQ217496.1 - 2, 3 381
FQ218027.1 - 2, 3 382
FQ223624.1 - 2, 3 381
FQ223672.1 - 2, 3 378
FQ223774.1 - 2, 3 383
FQ223938.1 - 2, 3 383
FQ224156.1 - 2, 3 379
FQ224523.1 - 2, 3 382
FQ224856.1 - 2, 3 382
FQ224909.1 - 2, 3 382
FQ224921.1 - 2, 3 379
FQ224975.1 - 2, 3 382

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
heart development
response to hormone
oxygen transport
slow-twitch skeletal muscle fiber contraction
response to hydrogen peroxide
enucleate erythrocyte differentiation
brown fat cell differentiation
oxygen transporter activity
iron ion binding
oxygen binding
heme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0