This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MB SIRNA Assays ›
Gene Symbol:
Mb
Gene Name:
myoglobin
Gene Aliases:
-
Chromosome Location:
Chr.7: 118101633 - 118108864 on Build Rnor_6.0
UniGene ID:
Rn.40511
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s133115

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021588.2 NP_067599.1 1, 2 120
GenBank mRNA AF197916.1 - 1, 2 93
BC070511.1 - 1, 2 120 7103504
FQ214690.1 - 1, 2 152
FQ214898.1 - 1, 2 151
FQ214986.1 - 1, 2 152
FQ215026.1 - 1, 2 152
FQ215032.1 - 1, 2 151
FQ215043.1 - 1, 2 151
FQ215217.1 - 1, 2 150
FQ215441.1 - 1, 2 151
FQ215511.1 - 1, 2 150
FQ215553.1 - 1, 2 151
FQ215560.1 - 1, 2 150
FQ215564.1 - 1, 2 150
FQ215642.1 - 1, 2 161
FQ215802.1 - 1, 2 152
FQ215880.1 - 1, 2 152
FQ215982.1 - 1, 2 151
FQ216001.1 - 1, 2 150
FQ216029.1 - 1, 2 149
FQ216127.1 - 1, 2 149
FQ216280.1 - 1, 2 158
FQ216299.1 - 1, 2 152
FQ216306.1 - 1, 2 153
FQ216335.1 - 1, 2 152
FQ216380.1 - 1, 2 151
FQ216443.1 - 1, 2 150
FQ216504.1 - 1, 2 151
FQ216709.1 - 1, 2 152
FQ216731.1 - 1, 2 150
FQ216841.1 - 1, 2 149
FQ217450.1 - 1, 2 151
FQ217496.1 - 1, 2 150
FQ218027.1 - 1, 2 151
FQ223624.1 - 1, 2 150
FQ223672.1 - 1, 2 147
FQ223774.1 - 1, 2 152
FQ223938.1 - 1, 2 152
FQ224156.1 - 1, 2 148
FQ224523.1 - 1, 2 151
FQ224909.1 - 1, 2 151
FQ224921.1 - 1, 2 148
FQ224975.1 - 1, 2 151

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
heart development
response to hormone
oxygen transport
slow-twitch skeletal muscle fiber contraction
response to hydrogen peroxide
enucleate erythrocyte differentiation
brown fat cell differentiation
oxygen transporter activity
iron ion binding
oxygen binding
heme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0