This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NT5E SIRNA Assays ›
Gene Symbol:
Nt5e
Gene Name:
5' nucleotidase, ecto
Gene Aliases:
CD73, Nt5
Chromosome Location:
Chr.8: 95969002 - 96012733 on Build Rnor_6.0
UniGene ID:
Rn.40132
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132856

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nt5e
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021576.2 NP_067587.2 9 1719
GenBank mRNA BC081806.1 - 9 1749 7134340
BC098665.1 - 9 1719 7322840
J05214.1 - 9 2069

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

purine nucleotide biosynthetic process
AMP catabolic process
brain development
response to aluminum ion
dephosphorylation
adenosine metabolic process
adenosine biosynthetic process
positive regulation of lipid biosynthetic process
negative regulation of inflammatory response
nucleotide binding
ferrous iron binding
5'-nucleotidase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0