This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ESPN SIRNA Assays ›
Gene Symbol:
Espn
Gene Name:
espin
Gene Aliases:
Je
Chromosome Location:
Chr.5: 169293356 - 169331338 on Build Rnor_6.0
UniGene ID:
Rn.91373
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132706

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Espn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006239476.2 XP_006239538.1 12 2663
XM_006239479.2 XP_006239541.1 11 2600
XM_006239484.2 XP_006239546.1 3 1381
XM_008764375.1 XP_008762597.1 12 2672
XM_008764376.1 XP_008762598.1 12 2657
XM_008764377.1 XP_008762599.1 11 2624
XM_008764378.1 XP_008762600.1 11 2609
XM_008764379.1 XP_008762601.1 13 2582
XM_008764380.1 XP_008762602.1 12 2534
GenBank mRNA AF076856.1 - 3 642
AF540946.1 - 7 1354
AF540947.1 - 8 1381
AF540948.1 - 6 1126
AF540949.1 - 7 1153
AY587568.1 - 5 713
AY587569.1 - 4 803
U46007.1 - 12 2348

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
sensory perception of sound
locomotory behavior
parallel actin filament bundle assembly
actin filament bundle assembly
positive regulation of filopodium assembly
negative regulation of cytoskeleton organization
SH3 domain binding
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0