This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPT2 SIRNA Assays ›
Gene Symbol:
Ppt2
Gene Name:
palmitoyl-protein thioesterase 2
Gene Aliases:
-
Chromosome Location:
Chr.20: 4381455 - 4401983 on Build Rnor_6.0
UniGene ID:
Rn.127781
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132656

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppt2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019367.2 NP_062240.1 4 515
XM_006256017.1 XP_006256079.1 4 830
XM_006256018.2 XP_006256080.1 4 1652
XM_006256019.2 XP_006256081.1 4 1385
XM_006256020.2 XP_006256082.1 4 1240
XM_006256022.2 XP_006256084.1 4 1199
XM_006256023.2 XP_006256085.1 4 673
XM_008772762.1 XP_008770984.1 4 2139
GenBank mRNA AF061971.1 - 4 447
AF067790.1 - 4 487
BC062023.1 - 4 477 5623515
FQ234132.1 - 4 457

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
macromolecule depalmitoylation
palmitoyl-(protein) hydrolase activity
thiolester hydrolase activity
palmitoyl hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0