This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDPN SIRNA Assays ›
Gene Symbol:
Pdpn
Gene Name:
podoplanin
Gene Aliases:
E11, Gp38, OTS-8, RTI40, T1-alpha
Chromosome Location:
Chr.5: 161947137 - 161981441 on Build Rnor_6.0
UniGene ID:
Rn.794
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132628

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pdpn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019358.1 NP_062231.1 4 607
XM_006239338.2 XP_006239400.1 4 607
GenBank mRNA BC072492.1 - 4 563 7097186
U07797.1 - 4 607
U32115.1 - 4 535
U96449.1 - 4 387

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
lymphangiogenesis
prostaglandin metabolic process
water transport
amino acid transport
inflammatory response
cell adhesion
signal transduction
nervous system development
cell proliferation
regulation of cell shape
positive regulation of platelet activation
folic acid transport
single organismal cell-cell adhesion
lung development
positive regulation of cell migration
tube morphogenesis
lung alveolus development
positive regulation of cellular component movement
response to hyperoxia
regulation of G1/S transition of mitotic cell cycle
molecular_function
water transmembrane transporter activity
folic acid transporter activity
amino acid transmembrane transporter activity
water channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0