This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLK SIRNA Assays ›
Gene Symbol:
Slk
Gene Name:
STE20-like kinase
Gene Aliases:
Stk2
Chromosome Location:
Chr.1: 267359769 - 267415735 on Build Rnor_6.0
UniGene ID:
Rn.228809
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132605

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slk
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019349.2 NP_062222.2 14 3155
XM_006231558.2 XP_006231620.1 15 3286
XM_008760447.1 XP_008758669.1 16 4545

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
signal transduction by protein phosphorylation
positive regulation of histone phosphorylation
intracellular signal transduction
positive regulation of apoptotic process
protein autophosphorylation
cellular response to tumor necrosis factor
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0