This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RGS12 SIRNA Assays ›
Gene Symbol:
Rgs12
Gene Name:
regulator of G-protein signaling 12
Gene Aliases:
-
Chromosome Location:
Chr.14: 80975239 - 81069517 on Build Rnor_6.0
UniGene ID:
Rn.9796
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132567

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rgs12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019339.1 NP_062212.1 1 466
XM_006251351.2 XP_006251413.1 2 598
XM_006251352.2 XP_006251414.1 2 602
XM_006251353.2 XP_006251415.1 2 1122
XM_008770350.1 XP_008768572.1 2 1135
GenBank mRNA U92280.1 - 1 466

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular signal transduction
termination of G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
negative regulation of G-protein coupled receptor protein signaling pathway
regulation of catalytic activity
G-protein alpha-subunit binding
receptor signaling protein activity
GTPase activator activity
protein binding
GTPase regulator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0