This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BTG3 SIRNA Assays ›
Gene Symbol:
Btg3
Gene Name:
BTG family, member 3
Gene Aliases:
-
Chromosome Location:
Chr.11: 16873642 - 16889100 on Build Rnor_6.0
UniGene ID:
Rn.8897
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132414

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Btg3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019290.1 NP_062163.1 3, 4 358
GenBank mRNA AF087037.1 - 3, 4 358

Back To Top

More Information


Gene Ontology Categories:

Function(s)

response to oxidative stress
negative regulation of cell proliferation
negative regulation of mitotic cell cycle

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0