This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GSK3A SIRNA Assays ›
Gene Symbol:
Gsk3a
Gene Name:
glycogen synthase kinase 3 alpha
Gene Aliases:
-
Chromosome Location:
Chr.1: 82097244 - 82108203 on Build Rnor_6.0
UniGene ID:
Rn.36807
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132334

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gsk3a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017344.1 NP_059040.1 2 485
XM_008758972.1 XP_008757194.1 2 473
GenBank mRNA X53427.1 - 2 485

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of systemic arterial blood pressure
cardiac left ventricle morphogenesis
glycogen metabolic process
regulation of gene expression by genetic imprinting
protein phosphorylation
signal transduction
nervous system development
insulin receptor signaling pathway
positive regulation of peptidyl-threonine phosphorylation
Wnt signaling pathway
cell migration
positive regulation of cAMP biosynthetic process
negative regulation of TOR signaling
cellular response to insulin stimulus
positive regulation of peptidyl-serine phosphorylation
cellular response to interleukin-3
proteasome-mediated ubiquitin-dependent protein catabolic process
hypermethylation of CpG island
positive regulation of heart contraction
positive regulation of transcription from RNA polymerase II promoter
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
negative regulation of transferase activity
negative regulation of cell growth involved in cardiac muscle cell development
cellular response to lithium ion
cellular response to organic cyclic compound
positive regulation of adrenergic receptor signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
positive regulation of glycogen (starch) synthase activity
protein kinase activity
protein serine/threonine kinase activity
ATP binding
protein kinase A catalytic subunit binding
tau-protein kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0