This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAB3IP SIRNA Assays ›
Gene Symbol:
Rab3ip
Gene Name:
RAB3A interacting protein
Gene Aliases:
RABIN3
Chromosome Location:
Chr.7: 59927486 - 59971516 on Build Rnor_6.0
UniGene ID:
Rn.31889
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132189

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rab3ip
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017313.3 NP_059009.2 2, 3 544
XM_006241357.2 XP_006241419.1 3, 4 713
XM_006241358.2 XP_006241420.1 3, 4 718
XM_006241359.2 XP_006241421.1 3, 4 1722
XM_006241360.2 XP_006241422.1 3, 4 708
GenBank mRNA BC127500.1 - 2, 3 547 7457420
U19181.1 - 2, 3 537

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK203553, AK152122, S79539.

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to membrane
protein transport
protein localization to organelle
cilium assembly
protein binding
Rab guanyl-nucleotide exchange factor activity
GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0