This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GCLM SIRNA Assays ›
Gene Symbol:
Gclm
Gene Name:
glutamate cysteine ligase, modifier subunit
Gene Aliases:
Glclr
Chromosome Location:
Chr.2: 225827504 - 225847876 on Build Rnor_6.0
UniGene ID:
Rn.2460
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132139

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gclm
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017305.2 NP_059001.1 5 597
GenBank mRNA BC078867.1 - 5 597 7110139
DQ266366.1 - 4 339
L22191.1 - 5 535
S65555.1 - 5 535

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cysteine metabolic process
glutamate metabolic process
glutathione metabolic process
glutathione biosynthetic process
response to oxidative stress
apoptotic mitochondrial changes
positive regulation of glutamate-cysteine ligase activity
response to drug
negative regulation of neuron apoptotic process
regulation of blood vessel size
response to nitrosative stress
regulation of mitochondrial depolarization
negative regulation of extrinsic apoptotic signaling pathway
glutamate-cysteine ligase activity
enzyme regulator activity
glutamate-cysteine ligase catalytic subunit binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0