This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CACNA1G SIRNA Assays ›
Gene Symbol:
Cacna1g
Gene Name:
calcium channel, voltage-dependent, T type, alpha 1G subunit
Gene Aliases:
-
Chromosome Location:
Chr.10: 82129071 - 82197863 on Build Rnor_6.0
UniGene ID:
Rn.86960
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132094

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cacna1g
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031601.3 NP_113789.2 8 1733
XM_008767985.1 XP_008766207.1 8 1751
XM_008767986.1 XP_008766208.1 8 1751
XM_008767987.1 XP_008766209.1 8 1751
XM_008767988.1 XP_008766210.1 8 1751
XM_008767989.1 XP_008766211.1 8 1751
XM_008767990.1 XP_008766212.1 8 1751
XM_008767991.1 XP_008766213.1 8 1751
XM_008767992.1 XP_008766214.1 8 1751
XM_008767993.1 XP_008766215.1 8 1751
XM_008767994.1 XP_008766216.1 8 1751
XM_008767995.1 XP_008766217.1 8 1751
XM_008767996.1 XP_008766218.1 8 1751
XM_008767997.1 XP_008766219.1 8 1751
XM_008767998.1 XP_008766220.1 8 1751
XM_008767999.1 XP_008766221.1 8 1751
XM_008768000.1 XP_008766222.1 8 1700
XM_008768001.1 XP_008766223.1 8 1751
GenBank mRNA AF027984.1 - 8 1733
AF125161.1 - 8 1408
AF290212.2 - 8 1364
EF116282.1 - 8 1201
EF116283.2 - 3 325
FJ227500.1 - 3 274

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001177888, NM_001177890, NM_009783, NM_001112813, DQ317412, and others.

Gene Ontology Categories:

Function(s) Process(es)

action potential
regulation of heart rate
positive regulation of cytosolic calcium ion concentration
chemical synaptic transmission
response to nickel cation
artery smooth muscle contraction
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of calcium ion-dependent exocytosis
regulation of calcium ion transport
regulation of atrial cardiac muscle cell membrane depolarization
calcium ion transport into cytosol
calcium ion import
calcium ion transmembrane transport
cellular response to dexamethasone stimulus
membrane depolarization during action potential
voltage-gated calcium channel activity
low voltage-gated calcium channel activity
scaffold protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0