This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PLA2G2A SIRNA Assays ›
Gene Symbol:
Pla2g2a
Gene Name:
phospholipase A2, group IIA (platelets, synovial fluid)
Gene Aliases:
sPLA2
Chromosome Location:
Chr.5: 157282650 - 157285295 on Build Rnor_6.0
UniGene ID:
Rn.11346
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s132032

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pla2g2a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031598.3 NP_113786.3 5 434
XM_006239146.2 XP_006239208.1 6 600
GenBank mRNA AF365363.1 - 4 378
AY651027.1 - 4 378
D00523.1 - 5 418
FQ220554.1 - 3 898
FQ227448.1 - 5 434
FQ234749.1 - 6 532
M25148.1 - 5 434
X74364.1 - 3 314

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phospholipid metabolic process
negative regulation of cell proliferation
lipid catabolic process
somatic stem cell population maintenance
regulation of cell proliferation
phosphatidic acid metabolic process
regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
phospholipase A2 activity
calcium ion binding
phospholipid binding
calcium-dependent phospholipase A2 activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0