This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNAPC1 SIRNA Assays ›
Gene Symbol:
SNAPC1
Gene Name:
small nuclear RNA activating complex polypeptide 1
Gene Aliases:
PTFgamma, SNAP43
Chromosome Location:
Chr.14: 61762357 - 61796428 on Build GRCh38
UniGene ID:
Hs.179312
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s13192

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNAPC1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003082.3 NP_003073.1 7 886
GenBank mRNA AK314933.1 - 7 886
BC014984.1 - 7 794 4872279
BC019038.2 - 7 790 4590014
U44754.1 - 7 794
Z47542.1 - 7 806

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
snRNA transcription from RNA polymerase II promoter
snRNA transcription from RNA polymerase III promoter
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0